Skip to Content
MilliporeSigma
Ethanol Assay Kit sufficient for 500 colorimetric tests

Ethanol Assay Kit

Ethanol Assay Kit sufficient for 500 colorimetric tests

Products

Product Category
Brand
Detection Method
Quality Segment
Application
Relevant Disease
Showing 1-2 of 2