Skip to Content
MilliporeSigma
IL-15 human Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

IL-15 human

IL-15 human Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Synonym(s):
IL-T

Products

Product Category
Brand
Biological Source
Color
Gene Alias
Physical Form
Recombinant Host
Technique