Skip to Content
MilliporeSigma
HumanKine® IL-1 beta, Human Recombinant Xeno-Free

IL-1 beta human

HumanKine® IL-1 beta, Human Recombinant Xeno-Free
Synonym(s):
Catabolin, Endogenous Pyrogen (EP),, Leukocyte Endogenous Mediator (LEM),, Lymphocyte-activating factor (LAF), Mononuclear Cell Factor (MCF)

Products

Product Category
Brand
Biological Source
Color
Gene Alias
Physical Form
Recombinant Host
Research Area
Quality Segment
Technique