Skip to Content
MilliporeSigma
Iron Test colorimetric, 0.1-50 mg/L (Fe), for use with MCOLORTEST®

Iron Test Kit, colorimetric

Iron Test colorimetric, 0.1-50 mg/L (Fe), for use with MCOLORTEST®

Products

Product Category
Brand
Analyte Suitability
Detection Method
Quality Segment