Skip to Content
US
EN
Back
Isoliquiritin
Synonym(s):
2′,4,4′-Trihydroxychalcone 4-glucoside, Isoliquiritoside
Empirical Formula (Hill Notation):
C
21
H
22
O
9
CAS Number:
5041-81-6
Molecular Weight:
418.39
Products
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
SMB01045
Isoliquiritin,
≥90% (LC/MS-ELSD)
Expand
View Pricing
PHL80481
Isoliquiritin,
phyproof
®
Reference Substance
Expand
View Pricing