
IWP-2

Synonym(s):
N-(6-Methyl-2-benzothiazolyl)-2-[(3,4,6,7-tetrahydro-4-oxo-3-phenylthieno[3,2-d]pyrimidin-2-yl)thio]-acetamide, Wnt Antagonist II, IWP-2, InSolution Wnt Antagonist II, IWP-2
Empirical Formula (Hill Notation):
C22H18N4O2S3
CAS Number:
Molecular Weight:
466.60
Products
Shipping
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-3 of 3