MilliporeSigma
Kolliphor® P 188

Kolliphor® P 188

Kolliphor® P 188
Synonyms:
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)
CAS Number:

Products

Shipping
Product Category
Brand
pH Value
Physical Form
Agency
Chemical Composition
Quality Segment
Application

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-2 of 2
Compare
Product Number
Description
Pricing