MilliporeSigma
Kolliphor® P 407 oxyethylene 71.5-74.9 %

Kolliphor® P 407

Kolliphor® P 407 oxyethylene 71.5-74.9 %
Synonyms:
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)
CAS Number:

Products

Shipping
Product Category
Brand
pH Value
Chemical Composition

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-1 of 1
Compare
Product Number
Description
Pricing