Skip to Content
MilliporeSigma
Manganese Standard for ICP TraceCERT®, 10 g/L Mn in nitric acid (nominal concentration)

Manganese Standard for ICP

Manganese Standard for ICP TraceCERT®, 10 g/L Mn in nitric acid (nominal concentration)

Products

Shipping
Product Category
Brand
Product Line
Format
Element
Quality Segment
Application
Special Grade
Technique