Skip to Content
MilliporeSigma
MHY1485 ≥95% (HPLC)

MHY1485

MHY1485 ≥95% (HPLC)
Synonym(s):
mTOR Activator, MHY1485, 4,6-Di-4-morpholinyl-N-(4-nitrophenyl)-1,3,5-triazin-2-amine
Empirical Formula (Hill Notation):
C17H21N7O4
CAS Number:
Molecular Weight:
387.39

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing