MilliporeSigma
vibrant-m

Monoclonal Anti-CD73 antibody produced in mouse

vibrant-m

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique

Products

Showing 1-2 of 2