Skip to Content
MilliporeSigma
Monoclonal Anti-FYN antibody produced in mouse clone 1A3, purified immunoglobulin, buffered aqueous solution

Monoclonal Anti-FYN antibody produced in mouse

Monoclonal Anti-FYN antibody produced in mouse clone 1A3, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-FYN oncogene related to SRC, FGR, YES, Anti-MGC45350, Anti-SLK, Anti-SYN

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique