
Multimedia Durapore® 0.5/0.2/0.22 µm, Opticap® XL Capsule

Synonym(s):
Multimedia Durapore® 0.5/0.2/0.22 µm, Opticap® XL Capsule
Products
Product Category
Brand
Product Line
Sterilization Compatibility
Agency
Length (cm)
Length (in)
Quality Segment
Hydrophobicity
Housing Material
Support Material
Pore size (µm)
Sterility
Technique
Showing 1-1 of 1