Skip to Content
MilliporeSigma
n-Decane for synthesis

n-Decane

n-Decane for synthesis
Synonym(s):
n-Decane
Linear Formula:
CH3(CH2)8CH3
CAS Number:
Molecular Weight:
142.28

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing