Nitric acid fuming 100%
Synonym(s):
Nitric acid solution
Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
Products
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing