Paraplast Plus®
Synonym(s):
Paraffin – polyisobutylene mixture
CAS Number:
EC Number:
Products
Product Category
Brand
Melting Point (°C)
Physical Form
Quality Segment
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing