Skip to Content
MilliporeSigma
Rat IL-1 β ELISA Kit for cell and tissue lysates

Rat IL-1 β ELISA Kit

Rat IL-1 β ELISA Kit for cell and tissue lysates

Products

Product Category
Brand
Gene Alias
Detection Method
Package Size
Species Reactivity
Technique
Showing 1-2 of 2