
SB225002

Synonym(s):
N-(2-Hydroxy-4-nitrophenyl)-N′-(2-bromophenyl)urea, SB-225002, SB 225002
Empirical Formula (Hill Notation):
C13H10BrN3O4
CAS Number:
Molecular Weight:
352.14
Products
Shipping
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2