MilliporeSigma
vibrant-m

Small Particle GCB Adsorbent

vibrant-m
EC Number:

Products

Product Category
Brand
Physical Form
Separation Technique
Quality Segment
Package Size
Particle Size (µm)
Pore Size (Å)
Technique