Skip to Content
US
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium hydroxide solution
Products
Shipping
Ships Today (1)
Brand
SAFC (6)
Sigma-Aldrich (1)
Supelco (1)
Physical Form
Product Line
Quality Segment
Application
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
2105
Sodium hydroxide solution,
0.1 N
Expand
View Pricing
1.00238
Sodium hydroxide solution,
50%, EMPROVE
®
EXPERT
Expand
View Pricing
1.00232
Sodium hydroxide solution,
10%, EMPROVE
®
EXPERT
Expand
View Pricing
1.05596
Sodium hydroxide solution,
about 36%, for analysis EMSURE
®
Expand
View Pricing
1.00221
Sodium hydroxide solution,
10 M, SUITABLE FOR CLEANING IN PLACE
Expand
View Pricing
4.80659
Sodium hydroxide solution,
25% (low iron), EMPROVE
®
EXPERT
Expand
View Pricing
4.80763
Sodium hydroxide solution,
8 M, EMPROVE
®
EXPERT
Expand
View Pricing
1.00233
Sodium hydroxide solution,
0.25 M, EMPROVE
®
EXPERT
Expand
View Pricing