MilliporeSigma
Manganese(II) chloride 0.1 M solution

Manganese(II) chloride solution

Manganese(II) chloride 0.1 M solution
Synonyms:
Manganese(II) chloride solution
Linear Formula:
MnCl2
CAS Number:
Molecular Weight:
125.84

Products

Shipping
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Sterilization Method

Products

Showing 1-2 of 2