MilliporeSigma
Methanol solution NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.

Hemacolor® Rapid staining of blood smear

Methanol solution NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Synonyms:
Methanol, Methyl alcohol, Methanol solution
Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique