MilliporeSigma
Hydrogen chloride solution c(HCl) = 3.571 mol/l (1/0.28 N), Titripur®

Hydrochloric acid solution

Hydrogen chloride solution c(HCl) = 3.571 mol/l (1/0.28 N), Titripur®
Synonym(s):
Hydrochloric acid solution, Hydrogen chloride solution, Hydrochloric acid, Hydrochloric acid standard solution
Linear Formula:
HCl
CAS Number:
Molecular Weight:
36.46

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Package Size
Reaction Type
Technique
Showing 1-20 of 35
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2