MilliporeSigma
Phenol (90% solution in water) for synthesis

Phenol solution

Phenol (90% solution in water) for synthesis
Synonyms:
Phenol solution
Empirical Formula (Hill Notation):
C6H6O
CAS Number:
Molecular Weight:
94.11

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique