MilliporeSigma
vibrant-m

Sodium hyaluronate for IR Identification

vibrant-m
Synonym(s):
Hyaluronic acid sodium salt
CAS Number:

Products

Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1