MilliporeSigma
L-Leucine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

L-Leucine

L-Leucine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(S)-2-Amino-4-methylpentanoic acid, L-Leucine
Linear Formula:
(CH3)2CHCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
EC Number:

Products

Shipping
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique