MilliporeSigma
L-Aspartic acid phyproof® Reference Substance

L-Aspartic acid

L-Aspartic acid phyproof® Reference Substance
Synonym(s):
(S)-(+)-Aminosuccinic acid, (S)-Aminobutanedioic acid, L-Aspartic acid
Linear Formula:
HO2CCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
133.10

Products

Shipping
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique