MilliporeSigma
Iodomethane solution certified reference material, 2000 μg/mL in methanol: water (4:1)

Iodomethane

Iodomethane solution certified reference material, 2000 μg/mL in methanol: water (4:1)
Synonym(s):
Iodomethane, Methyl iodide
Empirical Formula (Hill Notation):
CH3I
CAS Number:
Molecular Weight:
141.94

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique