MilliporeSigma
Pantothenate for peak identification European Pharmacopoeia (EP) Reference Standard

Calcium-d-pantothenate

Pantothenate for peak identification European Pharmacopoeia (EP) Reference Standard
Synonyms:
D-Pantothenic acid hemicalcium salt, (R)-(+)-N-(2,4-Dihydroxy-3,3-dimethyl-1-oxobutyl)-β-alanine hemicalcium salt, Calcium D-pantothenate, Vitamin B5
Linear Formula:
HOCH2C(CH3)2CH(OH)CONHCH2CH2CO2 ·1/2Ca
CAS Number:
Molecular Weight:
238.27

Products

Shipping
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique