Skip to Content
MilliporeSigma
Sulfur ICP standard traceable to SRM from NIST H2SO4 in H2O 10000 mg/l S Certipur®

Sulfur ICP standard

Sulfur ICP standard traceable to SRM from NIST H2SO4 in H2O 10000 mg/l S Certipur®

Products

Product Category
Brand
Physical Form
Product Line
Format
Element
Quality Segment
Application
Special Grade
Technique