MilliporeSigma
SUPELCOSIL™ LC-NH2 HPLC Column 5 μm particle size, L × I.D. 25 cm × 3 mm

SUPELCOSIL LC-NH2 HPLC Column

SUPELCOSIL™ LC-NH2 HPLC Column 5 μm particle size, L × I.D. 25 cm × 3 mm
EC Number:

Products

Shipping
Product Category
Brand
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique