MilliporeSigma
  • Hydroxysteroid (17β) dehydrogenase 1 expressed by Sertoli cells contributes to steroid synthesis and is required for male fertility.
Hydroxysteroid (17β) dehydrogenase 1 expressed by Sertoli cells contributes to steroid synthesis and is required for male fertility.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-02-07)
Janne Hakkarainen, Fu-Ping Zhang, Heli Jokela, Artur Mayerhofer, Rüdiger Behr, Sheyla Cisneros-Montalvo, Mirja Nurmio, Jorma Toppari, Claes Ohlsson, Noora Kotaja, Petra Sipilä, Matti Poutanen
ABSTRACT
The pituitary gonadotrophins and testosterone are the main hormonal regulators of spermatogenesis, but estradiol is also known to play a role in the process. The hormonal responses in the testis are partially mediated by somatic Sertoli cells that provide nutritional and physical support for differentiating male germ cells. Hydroxysteroid (17β) dehydrogenase 1 (HSD17B1) is a steroidogenic enzyme that especially catalyzes the conversion of low potent 17keto-steroids to highly potent 17β-hydroxysteroids. In this study, we show that Hsd17b1 is highly expressed in Sertoli cells of fetal and newborn mice, and HSD17B1 knockout males present with disrupted spermatogenesis with major defects, particularly in the head shape of elongating spermatids. The cell-cell junctions between Sertoli cells and germ cells were disrupted in the HSD17B1 knockout mice. This resulted in complications in the orientation of elongating spermatids in the seminiferous epithelium, reduced sperm production, and morphologically abnormal spermatozoa. We also showed that the Sertoli cell-expressed HSD17B1 participates in testicular steroid synthesis, evidenced by a compensatory up-regulation of HSD17B3 in Leydig cells. These results revealed a novel role for HSD17B1 in the control of spermatogenesis and male fertility, and that Sertoli cells significantly contribute to steroid synthesis in the testis.-Hakkarainen, J., Zhang, F.-P., Jokela, H., Mayerhofer, A., Behr, R., Cisneros-Montalvo, S., Nurmio, M., Toppari, J., Ohlsson, C., Kotaja, N., Sipilä, P., Poutanen, M. Hydroxysteroid (17β) dehydrogenase 1 expressed by Sertoli cells contributes to steroid synthesis and is required for male fertility.
Sigma-Aldrich
Epoxy Embedding Medium kit, embedding resin for electron microscopy
Sigma-Aldrich
Triton X-100, laboratory grade