MilliporeSigma
  • Factors involved in specific transcription by mammalian RNA polymerase II. Factors IIE and IIF independently interact with RNA polymerase II.
Factors involved in specific transcription by mammalian RNA polymerase II. Factors IIE and IIF independently interact with RNA polymerase II.
The Journal of biological chemistry (1989-05-25)
O Flores, E Maldonado, D Reinberg
ABSTRACT
The purification and characterization of transcription factor IIF (TFIIF), a factor required for transcription by the RNA polymerase II machinery, is described. TFIIF was isolated from the previously described IIE protein fraction. TFIIF enters into the transcription cycle via a preinitiation complex, and it is required for the formation of a complex capable of initiating transcription in the presence of heparin concentrations that inhibit the action of a free factor. TFIIF and TFIIE independently interacted with purified RNA polymerase II. TFIIF and TFIIE were both required for transcription of several class II promoters, including a promoter that lacks the conserved TATA box. Interestingly TFIIF was absolutely required for the formation of a preinitiation complex; however, it also affected the elongation phase of the transcription cycle. TFIIF, together with the previously described elongation factor TFIIS, was required for efficient elongation.
Sigma-Aldrich
TFIIF (RAP74 subunit) human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)