MilliporeSigma
Duck RIG-I restricts duck enteritis virus infection.
Veterinary microbiology (2019-03-05)
Hong Huo, Yue Wang, Dongfang Wang, Yiping Wang, Xiaohan Chen, Lili Zhao, Hongyan Chen
ABSTRACT
Retinoic acid-inducible gene I (RIG-I) is a nucleic acid sensor that plays a key role in host antiviral defenses. Duck viral enteritis (DEV) is a DNA virus that causes significant economic losses to the poultry industry worldwide. Although RIG-I is known to be involved in a common antiviral signaling pathway triggered by RNA viruses, its role in DEV infection remains unclear. In this study, we demonstrated that DEV infection increased the expression levels of interferon β (IFN-β) and RIG-I in ducks both in vivo and in vitro. Furthermore, overexpression of duck RIG-I significantly upregulated the expression of interferon-stimulated genes, including myxovirus resistance protein (Mx), Interferon-induced oligodenylate synthetase-like (OASL) and IFN-β. We therefore used overexpression and knockdown methods to determine if RIG-I affected DEV infection in ducks. Viral infection was inhibited by RIG-I, and enhanced by knockdown of RIG-I expression using small interfering RNA. RIG-I overexpression also activated signal transducer and activator of transcription 1 (STAT1), as a member of the JAK-STAT family. The combined results following STAT1 knockdown and RIG-I overexpression suggested that the antiviral activity of RIG-I was STAT1-dependent. Overall, these findings indicate that RIG-I effectively restricts DEV replication and may play a vital role in the host immune response to DEV infection in ducks.