MilliporeSigma
  • The long-acting beta2-agonist salmeterol xinafoate: effects on airway inflammation in asthma.
The long-acting beta2-agonist salmeterol xinafoate: effects on airway inflammation in asthma.
The European respiratory journal (1999-10-09)
J A Roberts, P Bradding, K M Britten, A F Walls, S Wilson, C Gratziou, S T Holgate, P H Howarth
ABSTRACT
Salmeterol xinafoate is an inhaled long-acting beta2-adrenoceptor agonist recently introduced for the treatment of asthma. Both in vitro and animal studies suggest that it may have anti-inflammatory activities of benefit in this disease. To assess this directly, the effects of 6 weeks' treatment with salmeterol on indices of clinical activity, airway dysfunction and inflammation in subjects with stable atopic asthma were investigated. In a double blind study, asthmatic patients were randomized to 6 weeks' treatment with either salmeterol 50 microg twice daily (n=14) or placebo (n=12). They underwent bronchoscopy with bronchoalveolar lavage (BAL) and bronchial biopsy immediately before starting treatment and again after 6 weeks. Treatment with salmeterol improved clinical indices of asthma activity, but there were no changes in BAL differential cell counts or mediator levels, and no change in T-cell numbers or activation status. In the biopsy specimens there were no changes in numbers of inflammatory cells, sub-basement membrane collagen deposition or mast cell degranulation. Regular treatment with salmeterol improves clinical indices of asthma but has no effect on the underlying inflammatory process. These findings strengthen guideline recommendations that long-acting beta2-agonists should not be prescribed as sole antiasthma medication.
Sigma-Aldrich
IgG1, Kappa from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution