MilliporeSigma
  • Role of nucleoside transport inhibition and endogenous adenosine in prevention of catecholamine induced death in rabbits.
Role of nucleoside transport inhibition and endogenous adenosine in prevention of catecholamine induced death in rabbits.
Cardiovascular research (1993-01-01)
H Van Belle, K Ver Donck, W Verheyen
ABSTRACT
R 75,231, a potent and specific nucleoside transport inhibitor, largely prevents cardiac damage and death in catecholamine challenged rabbits. The major biochemical effect of nucleoside transport inhibition in ischaemic and reperfused myocardium is a prolonged accumulation of adenosine. The cardioprotection by R 75,231 may be explained if it can be shown that endogenous adenosine plays a role in catecholamine cardiotoxicity and if nucleoside transport inhibition is required for the cardioprotective effect of R 75,231. Several groups of rabbits were infused with catecholamines until death. Changes in survival with time of infusion by coinfusion of aminophylline and/or treatment with R 75,231 and its two stereoenantiomers were assessed. Treatment with R 75,231 postponed the time to reach 50% mortality threefold after challenge with adrenaline or noradrenaline. Draflazine, the (-)-enantiomer of R 75,231, was also effective, whereas the (+)-enantiomer, which is devoid of any effect on the transporter, was not cardioprotective. The cardioprotective effect of R 75,231 was dependent on the extent and duration of ex vivo inhibition of the transporter in blood. Co-infusion of aminophylline with adrenaline significantly accelerated the rate of mortality. Nucleoside transport inhibition is the major, if not the only, determinant for efficacy of R 75,231 and draflazine as cardioprotective agents. Taken together with the evidence for a role of endogenous adenosine, the benefit from nucleoside transport inhibition in this model may be the result of prolonged accumulation of endogenous adenosine.
Sigma-Aldrich
Draflazine, ≥98% (HPLC)