MilliporeSigma
  • Angiogenesis and host immune response contribute to the aggressive character of non-melanoma skin cancers in renal transplant recipients.
Angiogenesis and host immune response contribute to the aggressive character of non-melanoma skin cancers in renal transplant recipients.
Histopathology (2011-05-19)
Katarzyna Anna Mackenzie, Andrew Peter Miller, Barry David Hock, Jacqui Gardner, Jeremy William Simcock, Justin Allan Roake, Gabi Ursula Dachs, Bridget Anne Robinson, Margaret Jane Currie
ABSTRACT
The aim of this study was to determine the contribution of tumour angiogenesis to the aggressive growth of non-melanoma skin cancers (NMSCs) in renal transplant recipients (RTRs). The study cohort included RTRs (n = 38) with formalin-fixed paraffin-embedded tumour samples available from first post-transplant NMSC (NMSC1) surgically excised at Christchurch Hospital, New Zealand, from 1997 to 2007. Comparable samples excised from immunocompetent individuals (ICIs) (n = 36) were selected to accommodate confounding factors. Markers of tumour angiogenesis were evaluated by immunohistochemistry, and analysed for associations with clinicopathological variables. As compared with ICIs, RTRs had a higher proportion of tumours with high microvessel density (P = 0.008), high proliferating capillary index (P < 0.0001) and low microvessel pericyte coverage index (P < 0.0001), and RTRs had a shorter cumulative second NMSC (NMSC2)-free interval (P < 0.0001). ICIs had a higher proportion of tumours with a 'marked' number of vascular endothelial growth factor (VEGF)-A-positive leukocytes than RTRs (P = 0.04), and RTRs with a 'moderate/marked' number of VEGF-A-positive leukocytes had longer cumulative NMSC2-free intervals than those with a 'minimum' number (P = 0.02). This study demonstrates increased tumour angiogenesis in NMSC in RTRs, and suggests a role for VEGF-A-positive peritumoural leukocytes in suppressing NMSC development.
Sigma-Aldrich
IgG1, Kappa from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution