MilliporeSigma
  • MYC Drives Temporal Evolution of Small Cell Lung Cancer Subtypes by Reprogramming Neuroendocrine Fate.
MYC Drives Temporal Evolution of Small Cell Lung Cancer Subtypes by Reprogramming Neuroendocrine Fate.
Cancer cell (2020-06-01)
Abbie S Ireland, Alexi M Micinski, David W Kastner, Bingqian Guo, Sarah J Wait, Kyle B Spainhower, Christopher C Conley, Opal S Chen, Matthew R Guthrie, Danny Soltero, Yi Qiao, Xiaomeng Huang, Szabolcs Tarapcsák, Siddhartha Devarakonda, Milind D Chalishazar, Jason Gertz, Justin C Moser, Gabor Marth, Sonam Puri, Benjamin L Witt, Benjamin T Spike, Trudy G Oliver
ABSTRACT
Small cell lung cancer (SCLC) is a neuroendocrine tumor treated clinically as a single disease with poor outcomes. Distinct SCLC molecular subtypes have been defined based on expression of ASCL1, NEUROD1, POU2F3, or YAP1. Here, we use mouse and human models with a time-series single-cell transcriptome analysis to reveal that MYC drives dynamic evolution of SCLC subtypes. In neuroendocrine cells, MYC activates Notch to dedifferentiate tumor cells, promoting a temporal shift in SCLC from ASCL1+ to NEUROD1+ to YAP1+ states. MYC alternatively promotes POU2F3+ tumors from a distinct cell type. Human SCLC exhibits intratumoral subtype heterogeneity, suggesting that this dynamic evolution occurs in patient tumors. These findings suggest that genetics, cell of origin, and tumor cell plasticity determine SCLC subtype.
Sigma-Aldrich
Anti-UCHL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-POU2F3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
Sigma-Aldrich
apo-Transferrin human, powder, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Anti-Calcitonin Gene Related Peptide antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein