MilliporeSigma
  • Developmental neurotoxicity of perfluorinated chemicals modeled in vitro.
Developmental neurotoxicity of perfluorinated chemicals modeled in vitro.
Environmental health perspectives (2008-06-19)
Theodore A Slotkin, Emiko A MacKillop, Ronald L Melnick, Kristina A Thayer, Frederic J Seidler
ABSTRACT
The widespread detection of perfluoroalkyl acids and their derivatives in wildlife and humans, and their entry into the immature brain, raise increasing concern about whether these agents might be developmental neurotoxicants. We evaluated perfluorooctane sulfonate (PFOS), perfluorooctanoic acid (PFOA), perfluorooctane sulfonamide (PFOSA), and perfluorobutane sulfonate (PFBS) in undifferentiated and differentiating PC12 cells, a neuronotypic line used to characterize neurotoxicity. We assessed inhibition of DNA synthesis, deficits in cell numbers and growth, oxidative stress, reduced cell viability, and shifts in differentiation toward or away from the dopamine (DA) and acetylcholine (ACh) neurotransmitter phenotypes. In general, the rank order of adverse effects was PFOSA > PFOS > PFBS approximately PFOA. However, superimposed on this scheme, the various agents differed in their underlying mechanisms and specific outcomes. Notably, PFOS promoted differentiation into the ACh phenotype at the expense of the DA phenotype, PFBS suppressed differentiation of both phenotypes, PFOSA enhanced differentiation of both, and PFOA had little or no effect on phenotypic specification. These findings indicate that all perfluorinated chemicals are not the same in their impact on neurodevelopment and that it is unlikely that there is one simple, shared mechanism by which they all produce their effects. Our results reinforce the potential for in vitro models to aid in the rapid and cost-effective screening for comparative effects among different chemicals in the same class and in relation to known developmental neurotoxicants.
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, from USDA approved countries, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, γ-irradiated by SER-TAIN process, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
Sigma-Aldrich
Horse Serum, USA origin, Donor herd, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, suitable for, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Horse serum, liquid
Sigma-Aldrich
Horse Serum, Donor Herd, USA origin, Heat inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Horse Serum, Donor herd, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
SAFC
Fetal Bovine Serum, Australia origin, γ-irradiated, suitable for cell culture, suitable for hybridoma