MilliporeSigma
  • Receptor tyrosine kinase EphA2 mediates thrombin-induced upregulation of ICAM-1 in endothelial cells in vitro.
Receptor tyrosine kinase EphA2 mediates thrombin-induced upregulation of ICAM-1 in endothelial cells in vitro.
Thrombosis research (2008-09-05)
Barden Chan, Vikas P Sukhatme
ABSTRACT
Thrombin potently induces endothelial inflammation. One of the responses is upregulation of adhesion molecules such as ICAM-1, resulting in enhanced leukocyte attachment to the endothelium. In this report, we examine the contribution of EphA2 in thrombin-induced expression of ICAM-1 in human umbilical vein endothelial cells (HUVECs). We showed that thrombin transiently induced tyrosine- phosphorylation of EphA2 in a Src-kinase dependent manner. This transactivation was mediated through PAR-1, because a PAR-1 specific agonistic peptide also transactivated EphA2. Expression knockdown of endogenous EphA2 by siRNAs blocked ICAM-1 upregulation and leukocyte/endothelium attachment induced by thrombin. Overexpression of exogenous mouse EphA2 rescued both ICAM-1 expression and leukocyte attachment induced by thrombin in endogenous EphA2-knockdown HUVECs. Mechanistically, we showed EphA2 knockdown suppressed thrombin-induced serine 536 phosphorylation of NFkappaB, an event critical of ICAM-1 transcriptional upregulation. Collectively, our results strongly suggest EphA2 is a necessary component for thrombin-induced ICAM-1 upregulation.
Sigma-Aldrich
Fetal Bovine Serum, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, from USDA approved countries, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, γ-irradiated by SER-TAIN process, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
SAFC
Fetal Bovine Serum, Australia origin, γ-irradiated, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, suitable for, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma