MilliporeSigma
  • Sodium butyrate activates Notch1 signaling, reduces tumor markers, and induces cell cycle arrest and apoptosis in pheochromocytoma.
Sodium butyrate activates Notch1 signaling, reduces tumor markers, and induces cell cycle arrest and apoptosis in pheochromocytoma.
American journal of translational research (2009-12-04)
Max A Cayo, Ashley K Cayo, Sarah M Jarjour, Herbert Chen
ABSTRACT
Pheochromocytoma is a neuroendocrine (NE) tumor of the adrenal medulla for which surgical resection is the only therapy. However, 10-46% of tumors are metastatic or have malignant features, and are often inoperable. Our lab has demonstrated the importance of the Notch1 signaling pathway in NE neoplasia, indicating that this pathway could be a target for emergent treatments in pheochromocytoma. It has recently become clear that histone deacetylase (HDAC) inhibitors influence Notch1 signaling. We hypothesized that the HDAC inhibitor Sodium Butyrate (NaB) might activate Notch1 in pheochromocytoma resulting in altered tumor cell proliferation. Pheochromocytoma (PC-12) cells were treated with increasing concentrations of NaB. MTT cellular proliferation assay was used to determine the effect of NaB on PC-12 cell growth. Expression of Notch1, NE markers, and cell cycle proteins was studied using Western analysis. Untreated PC-12 cells lack Notch1 activity. Treatment with NaB led to a dose-dependent induction of Notch1 signaling, reduction of NE markers ASCL1 and CgA, and a significant reduction in cellular proliferation. Levels of expression of cyclin D1, p21, cleaved PARP, and cleaved caspase 3 proteins indicated the presence of cell cycle arrest and apoptosis following NaB treatment. NaB activated Notch1 signaling, inhibited cellular proliferation, reduced NE markers, and induced cell cycle arrest and apoptosis in pheochromocytoma cells. This data indicates that activation of Notch1 signaling is a promising potential therapy or palliative measure for pheochromocytoma that warrants further investigation.
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Horse Serum, Donor herd, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Horse Serum, Donor Herd, USA origin, Heat inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Horse serum, liquid
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
SAFC
Fetal Bovine Serum, Australia origin, γ-irradiated, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, suitable for, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, from USDA approved countries, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, γ-irradiated by SER-TAIN process, sterile-filtered, suitable for cell culture