MilliporeSigma
  • Beneficial effects of a novel RAGE inhibitor on early diabetic retinopathy and tactile allodynia.
Beneficial effects of a novel RAGE inhibitor on early diabetic retinopathy and tactile allodynia.
Molecular vision (2011-12-16)
Guangyuan Li, Jie Tang, Yunpeng Du, Chieh Allen Lee, Timothy S Kern
ABSTRACT
The receptor for advanced glycation end products (RAGE) has been implicated in the pathogenesis of numerous complications of diabetes. We assessed the effect of a novel RAGE fusion protein inhibitor on retinal histopathology and nerve function, and on retinal inflammation and oxidative stress. C57BL/6J mice were made diabetic with streptozotocin, and some were given a RAGE fusion protein (10, 100, or 300 µg per mouse 3 times per week). Mice were sacrificed at 2 months and 10 months into the study to assess retinal vascular histopathology, accumulation of albumin in the neural retina, cell loss in the ganglion cell layer, and biochemical and physiologic abnormalities in the retina. Tactile allodynia (light touch) was measured on a paw of each animal at 2 months. Leukostasis, expression of the intercellular adhesion molecule-1 (ICAM-1), accumulation of albumin in the neural retina, and nitration of retinal proteins were significantly increased in the retinas of mice diabetic for 2 months. The number of degenerate retinal capillaries was significantly increased in mice diabetic for 10 months, compared to the nondiabetic controls. Diabetes also enhanced sensitivity of peripheral nerves to tactile allodynia. All three doses of the RAGE fusion protein inhibited capillary degeneration, accumulation of albumin in the neural retina, nitration of retinal proteins, and tactile allodynia, demonstrating that biologically meaningful levels of the drug reached the retina. RAGE inhibition did tend to inhibit diabetes-induced retinal leukostasis and ICAM-1 expression (previously postulated to be important in the pathogenesis of retinopathy), but these effects were not statistically significant for the use of the lower doses of the drug that normalized the vascular histopathology. Inhibition of RAGE blocked the development of important lesions of diabetic retinopathy, but these beneficial effects seemed not to be mediated via leukostasis. RAGE inhibition also blocked the development of sensory allodynia in diabetes. RAGE is an important therapeutic target to inhibit the development of vascular and neural complications of diabetes.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Anti-Nitrotyrosine Antibody, clone 1A6, clone 1A6, Upstate®, from mouse
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Periodic acid, ACS reagent, 99%
Sigma-Aldrich
Periodic acid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Periodic acid, puriss. p.a., ACS reagent, for oxidimetric titration, crystallized, ≥99.0% (RT)
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Sigma-Aldrich
Periodic acid, 99.999% trace metals basis
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Sigma-Aldrich
TWEEN® 20, viscous liquid, suitable for cell culture
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls
Sigma-Aldrich
TWEEN® 20, viscosity 250-450 mPa.s (25 °C)
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls
Sigma-Aldrich
Periodic acid, suitable for electrophoresis, ≥99%