MilliporeSigma
  • Identification of 3-(acylamino)azepan-2-ones as stable broad-spectrum chemokine inhibitors resistant to metabolism in vivo.
Identification of 3-(acylamino)azepan-2-ones as stable broad-spectrum chemokine inhibitors resistant to metabolism in vivo.
Journal of medicinal chemistry (2005-02-04)
David J Fox, Jill Reckless, Sibylle M Wilbert, Ian Greig, Stuart Warren, David J Grainger
ABSTRACT
3-(acylamino)glutarimides, a class of broad spectrum chemokine inhibitors, are rapidly hydrolyzed in serum, despite being stable in aqueous solution. Synthesis and high-performance liquid chromatography analysis of the proposed N-acyl-glutamate and -glutamine metabolites establish the enzyme-catalyzed breakdown pathways. In vitro assays suggest that despite their short half-life in vivo, the parent acylamino-glutarimides, not the ring-opened hydrolysis products, are the source of the antiinflammatory activity. Identification of this metabolic pathway has led to the development of 3-(acylamino)azepan-2-ones that are also broad spectrum chemokine inhibitors and act as stable, orally available powerful antiinflammatory agents in vivo with doses of 1 mg/kg.
Sigma-Aldrich
L-2-Aminoadipic acid, ≥98% (TLC)
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)