MilliporeSigma
  • Sublethal Paraquat Confers Multidrug Tolerance in Pseudomonas aeruginosa by Inducing Superoxide Dismutase Activity and Lowering Envelope Permeability.
Sublethal Paraquat Confers Multidrug Tolerance in Pseudomonas aeruginosa by Inducing Superoxide Dismutase Activity and Lowering Envelope Permeability.
Frontiers in microbiology (2020-10-27)
Dorival Martins, Geoffrey A McKay, Ann M English, Dao Nguyen
ABSTRACT
Stressors and environmental cues shape the physiological state of bacteria, and thus how they subsequently respond to antibiotic toxicity. To understand how superoxide stress can modulate survival to bactericidal antibiotics, we examined the effect of intracellular superoxide generators, paraquat and menadione, on stationary-phase antibiotic tolerance of the opportunistic pathogen, Pseudomonas aeruginosa. We tested how pre-challenge with sublethal paraquat and menadione alters the tolerance to ofloxacin and meropenem in wild-type P. aeruginosa and mutants lacking superoxide dismutase (SOD) activity (sodAB), the paraquat responsive regulator soxR, (p)ppGpp signaling (relA spoT mutant), or the alternative sigma factor rpoS. We confirmed that loss of SOD activity impairs ofloxacin and meropenem tolerance in stationary phase cells, and found that sublethal superoxide generators induce drug tolerance by stimulating SOD activity. This response is rapid, requires de novo protein synthesis, and is RpoS-dependent but does not require (p)ppGpp signaling nor SoxR. We further showed that pre-challenge with sublethal paraquat induces a SOD-dependent reduction in cell-envelope permeability and ofloxacin penetration. Our results highlight a novel mechanism of hormetic protection by superoxide generators, which may have important implications for stress-induced antibiotic tolerance in P. aeruginosa cells.
Sigma-Aldrich
Methyl viologen dichloride hydrate, 98%
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
Menadione, crystalline
Supelco
Activated charcoal, powder
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Xanthine Oxidase from bovine milk, Grade IV, ammonium sulfate suspension, ≥0.1 units/mg protein
Sigma-Aldrich
Gentamicin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Meropenem trihydrate, ≥98% (HPLC)