MilliporeSigma
  • Dietary valine levels affect growth, protein utilisation, immunity and antioxidant status in juvenile hybrid grouper (Epinephelus fuscoguttatus ♀ × Epinephelus lanceolatus ♂).
Dietary valine levels affect growth, protein utilisation, immunity and antioxidant status in juvenile hybrid grouper (Epinephelus fuscoguttatus ♀ × Epinephelus lanceolatus ♂).
The British journal of nutrition (2020-07-28)
Zhiyu Zhou, Xiaoyi Wu, Delbert M Gatlin, Xiao Wang, Wei Mu, Bo Ye, Lei Ma
ABSTRACT
A 6-week growth trial was conducted to evaluate the influences of dietary valine (Val) levels on growth, protein utilisation, immunity, antioxidant status and gut micromorphology of juvenile hybrid groupers. Seven isoenergetic, isoproteic and isolipidic diets were formulated to contain graded Val levels (1·21, 1·32, 1·45, 1·58, 1·69, 1·82 and 1·94 %, DM basis). Each experimental diet was hand-fed to triplicate groups of twelve hybrid grouper juveniles. Results showed that weight gain percentage (WG%), protein productive value, protein efficiency ratio, and feed efficiency were increased as dietary Val level increased, reaching a peak value at 1·58 % dietary Val. The quadratic regression analysis of WG% against dietary Val levels indicated that the optimum dietary Val requirement for hybrid groupers was estimated to be 1·56 %. Gut micromorphology and expression of growth hormone in pituitary, insulin-like growth factor 1, target of rapamycin and S6 kinase 1 in liver were significantly affected by dietary Val levels. In serum, fish fed 1·58 % dietary Val had higher superoxide dismutase, catalase, lysozyme activities and IgM concentrations than fish fed other dietary Val levels. Fish fed 1·58 % dietary Val had higher expression of NF-E2-related factor 2 in head kidney than fish fed other dietary Val levels. Generally, the optimum dietary Val requirement for maximal growth of hybrid groupers was estimated to be 1·56 % of DM, corresponding to 3·16 % of dietary protein, and dietary Val levels affected growth, protein utilisation, immunity and antioxidant status in hybrid groupers.
Sigma-Aldrich
Heparin ammonium salt from porcine intestinal mucosa, ≥140 USP units/mg