MilliporeSigma
  • Design and optimization of a hybrid process based on hollow-fiber membrane/coagulation for wastewater treatment.
Design and optimization of a hybrid process based on hollow-fiber membrane/coagulation for wastewater treatment.
Environmental science and pollution research international (2020-10-15)
Sajjad Alibeigi-Beni, Masoud Habibi Zare, Mahdi Pourafshari Chenar, Morteza Sadeghi, Saeed Shirazian
ABSTRACT
Treatment of textile wastewater using ultrafiltration membranes was carried out in this study. Since membrane fouling is a major operational problem that decreases the membrane separation efficiency, wastewater was treated with polyaluminum chloride (PACl) and alum (aluminum sulfate) as coagulant to decrease the fouling of ultrafiltration membranes. PACl was selected as the best coagulant in the experiments. Also, chitosan was used as coagulant aid upon developing the hybrid process. The obtained optimum dosage of PACl coagulant was 100 mg/L, and maximum turbidity and COD removal of 35% and 66% were attained, respectively. The pretreated wastewater by coagulation was sent to ultrafiltration process for further removal of turbidity and COD. Three ultrafiltration hollow-fiber membranes made of polypropylene (PP), polyvinylidene fluoride (PVDF), and polyethersulfone (PES) were applied in this study. In general, the filtration results were evaluated for two types of samples treated under coagulation and without treatment; the results were unfavorable for the second type. The effects of transmembrane pressure (TMP) and cross velocity on membranes performance were also investigated for process optimization. The obtained results showed that PVDF membrane had the highest flux and turbidity removal, whereas the PES membrane had the highest COD removal. Also, the results revealed that turbidity and COD removal by all membranes were decreased by increasing TMP and cross velocity.