MilliporeSigma
  • Electron Microscopic Detection of Proteins and Protein Complexes in Mammalian Cells Using APEX-tagged, Conditionally Stable Nanobodies.
Electron Microscopic Detection of Proteins and Protein Complexes in Mammalian Cells Using APEX-tagged, Conditionally Stable Nanobodies.
Bio-protocol (2018-11-20)
Thomas E Hall, James Rae, Nicholas Ariotti, Robert G Parton
ABSTRACT
The recent development of 3D electron microscopic techniques for cells and tissues has necessitated the development of new methods for the detection of proteins and protein-complexes in situ. The development of new genetic tags, such as the ascorbate peroxidase, APEX, for electron microscopic detection of tagged proteins has expanded the available toolbox and ushered in a new era in biological electron microscopy. Here, we describe methods for combining conditionally-stable nanobodies to fluorescent protein tags with APEX-based detection. These methods are compatible with detection of low levels of expression of fluorescently-tagged proteins and with detection of protein complexes using split GFP-based complementation methods. We describe a simple protocol for applying these methods to the electron microscopic detection of proteins and protein complexes in cultured cells.
Sigma-Aldrich
Sodium cacodylate trihydrate, ≥98%
Sigma-Aldrich
SIGMAFAST 3,3′-Diaminobenzidine tablets, tablet, To prepare 1 mL
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer