MilliporeSigma
Exposure to Neospora spp. and Besnoitia spp. in wildlife from Israel.
International journal for parasitology. Parasites and wildlife (2018-09-04)
Monica L Mazuz, Gema Alvarez-García, Roni King, Igor Savisky, Varda Shkap, Luis M Ortega-Mora, Daniel Gutiérrez-Expósito
ABSTRACT
Neosporosis and besnoitiosis, caused by cyst-forming protozoa Neospora caninum and Besnoitia besnoiti, respectively, are parasitic infestations of livestock in Israel. These parasites cause significant economic losses in cattle due to reproductive and productive disorders. Both parasites have been detected in several wild ruminant species throughout other regions of the world, while the existence of a sylvatic life cycle in Israel remains uncertain. Thus, a wide panel of 871 sera from two wild carnivores and nine wild ruminant species were tested. All sera were first analysed by MAT for an initial screening and positive samples were confirmed a posteriori by Western blot. Additionally, a complementary IFAT was used for the detection of antibodies against N. caninum. Neospora antibodies were present in six out of the 11 species investigated, whereas Besnoitia antibodies were undetected. Golden jackal, red fox, addax, Arabian oryx, Persian fallow deer, mouflon, mountain gazelle, Nubian ibex, scimitar horned oryx and water buffalo were seropositive against N. caninum infection by IFAT and/or MAT. Moreover, the presence of Neospora spp.-specific antibodies was confirmed by Western blot in golden jackal (6/189; 3.2%), red fox (1/75; 1.3%), Persian fallow deer (13/232; 5.6%), mouflon (1/15; 16.7%), Nubian ibex (22/55; 40%) and water buffalo (12/18; 66.7%). Addax (1/49) and water buffalo (1/18) were MAT-seropositive against B. besnoiti but were seronegative by Western blot. Hence, Neospora sylvatic cycle is present in Israel and may cross over to a domestic life cycle. In contrast, wildlife species investigated are unlikely to present a risk of transmitting Besnoitia to livestock in Israel.
Sigma-Aldrich
Protein A–Peroxidase from Staphylococcus aureus/horseradish, lyophilized powder