MilliporeSigma
  • Human alveolar progenitors generate dual lineage bronchioalveolar organoids.
Human alveolar progenitors generate dual lineage bronchioalveolar organoids.
Communications biology (2022-08-26)
Karen Hoffmann, Benedikt Obermayer, Katja Hönzke, Diana Fatykhova, Zeynep Demir, Anna Löwa, Luiz Gustavo Teixeira Alves, Emanuel Wyler, Elena Lopez-Rodriguez, Maren Mieth, Morris Baumgardt, Judith Hoppe, Theresa C Firsching, Mario Tönnies, Torsten T Bauer, Stephan Eggeling, Hong-Linh Tran, Paul Schneider, Jens Neudecker, Jens C Rückert, Achim D Gruber, Matthias Ochs, Markus Landthaler, Dieter Beule, Norbert Suttorp, Stefan Hippenstiel, Andreas C Hocke, Mirjana Kessler
ABSTRACT
Mechanisms of epithelial renewal in the alveolar compartment remain incompletely understood. To this end, we aimed to characterize alveolar progenitors. Single-cell RNA-sequencing (scRNA-seq) analysis of the HTII-280+/EpCAM+ population from adult human lung revealed subclusters enriched for adult stem cell signature (ASCS) genes. We found that alveolar progenitors in organoid culture in vitro show phenotypic lineage plasticity as they can yield alveolar or bronchial cell-type progeny. The direction of the differentiation is dependent on the presence of the GSK-3β inhibitor, CHIR99021. By RNA-seq profiling of GSK-3β knockdown organoids we identified additional candidate target genes of the inhibitor, among others FOXM1 and EGF. This gives evidence of Wnt pathway independent regulatory mechanisms of alveolar specification. Following influenza A virus (IAV) infection organoids showed a similar response as lung tissue explants which confirms their suitability for studies of sequelae of pathogen-host interaction.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Mucin MUC5AC Antibody, clone CLH2, clone CLH2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Prosurfactant Protein C (proSP-C) Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture