MilliporeSigma
  • EhRho6-mediated actin degradation in Entamoeba histolytica is associated with compromised pathogenicity.
EhRho6-mediated actin degradation in Entamoeba histolytica is associated with compromised pathogenicity.
Molecular microbiology (2022-03-25)
Anil Raj Narooka, Achala Apte, Pooja Yadav, Jimmy Rodriguez Murillo, Livia Goto-Silva, Magno Junqueira, Sunando Datta
ABSTRACT
Entamoeba histolytica causes amoebiasis which is a major health concern in developing countries. E. histolytica pathogenicity has been implicated to a large repertoire of small GTPases which switch between the inactive GDP bound state and the active GTP bound state with the help of guanine nucleotide exchange factors (GEFs) and GTPase activating protein (GAPs). Rho family of small GTPases are well known to modulate the actin cytoskeletal dynamics which plays a major role in E. histolytica pathogenicity. Here, we report an atypical amoebic RhoGEF, and its preferred substrate EhRho6, which, upon overexpression abrogated the pathogenic behavior of the amoeba such as adhesion to host cell, monolayer destruction, erythrophagocytosis, and formation of actin dots. A causative immunoblot analysis revealed actin degradation in the EhRho6 overexpressing trophozoites that could be inhibited by blocking the amoebic proteasomal pathway. A careful analysis of the results from a previously published transcriptomics study, in conjunction with our observations, led to the identification of a clade of Rho GTPases in this pathogenic amoeba which we hypothesize to have implications during the amoebic encystation.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Fibronectin human plasma, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture